Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus tribromide, 1.0M solution in dichloromethane, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
7789-60-8,75-09-2
Br3P
270.69
MFCD00011436
IPNPIHIZVLFAFP-UHFFFAOYSA-N
phosphorus tribromide,phosphorous tribromide,tribromophosphine,phosphorus iii bromide,phosphorous bromide,pbr3,phosphorus bromide,phosphorus bromide pbr3,unii-58r3866pua,tribro-mophosphine
24614
tribromophosphane
P(Br)(Br)Br
7789-60-8,75-09-2
Br3P
270.69
MFCD00011436
IPNPIHIZVLFAFP-UHFFFAOYSA-N
phosphorus tribromide,phosphorous tribromide,tribromophosphine,phosphorus iii bromide,phosphorous bromide,pbr3,phosphorus bromide,phosphorus bromide pbr3,unii-58r3866pua,tribro-mophosphine
24614
tribromophosphane
P(Br)(Br)Br
C-X bond formation reagent
Linear Formula PBr2
Merck Index 15, 7469
Formula Weight 270.69
Physical Form Liquid
Concentration 0.9 to 1.1M
Density 1.4880g/mL
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.488
Color Colorless to Yellow
Quantity 100mL
Chemical Name or Material Phosphorus tribromide, 1.0M Solution in Dichloromethane
Show More Show Less

GHS H Statement
Suspected of causing cancer if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 232-178-2