Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phosphorus(V) bromide, 95%

Click to view available options
Quantity:
25g,25g
100g,100g
500g,500g
7789-69-7
Br5P
430.494
MFCD00011437
QRKVRHZNLKTPGF-UHFFFAOYSA-N
phosphorus pentabromide,phosphorane, pentabromo,unii-3d9wis0bqw,pentabromophosphorane,phosphorus v bromide,3d9wis0bqw,pentabromo,phosphorpentabromid,phosphoruspentabromide,phosphorouspentabromide
62678
pentabromo-$l^{5}-phosphane
P(Br)(Br)(Br)(Br)Br
7789-69-7
Br5P
430.494
MFCD00011437
QRKVRHZNLKTPGF-UHFFFAOYSA-N
phosphorus pentabromide,phosphorane, pentabromo,unii-3d9wis0bqw,pentabromophosphorane,phosphorus v bromide,3d9wis0bqw,pentabromo,phosphorpentabromid,phosphoruspentabromide,phosphorouspentabromide
62678
pentabromo-$l^{5}-phosphane
P(Br)(Br)(Br)(Br)Br
Applications
Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Boiling Point 105°C to 107°C (decomposition),105° to 107°C (dec.)
Melting Point ~105°C (decomposition),≈105°C (dec.)
Odor Acrid,Acrid
Quantity 25g,25g
UN Number UN2691
Beilstein 3903332
Merck Index 14,7350
Formula Weight 430.5
Percent Purity 95%
Chemical Name or Material Phosphorus(V) bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHOSPHORUS PENTABROMIDE

TSCA : TSCA