Learn More
Phosphorus(V) chloride, 98%, Thermo Scientific Chemicals
Description
Phosphorus(V) chloride can be used for both oxidative and substitutive chlorinations, & for both organic and inorganic compounds. Dehydrating agent. Precursor to Vilsmeier reagent, lithium hexafluorophosphate electrolyite for lithium batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 162° (sublimation) |
Color | Yellow |
Physical Form | Solid |
Quantity | 4 x 500 g |
Assay Percent Range | 98% |
Solubility Information | Soluble in carbon disulfide and carbon tetrachloride. |
Formula Weight | 208.24 |
Odor | Pungent |
Sensitivity | Moisture sensitive |
Appearance | Yellowish crystals |
Show More |
Safety and Handling
Hazard Category | H302-H314-H330-H373 |
Precautionary Statement | P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | DOT Class: 8, Packaging Group: II |
EINECSNumber | 233-060-3 |
RTECSNumber | TB6125000 |
TSCA | Yes |
storageNote1 | Moisture Sensitive |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.