Phosphorus(V) oxide, ACS, 98% min, Alfa Aesar™

For Use As a Drying and Dehydrating Agent, As a Condensing Agent in organic Synthesis, in Sugar Refining

Manufacturer: Alfa Aesar 1052418

Quantity 50g

 View more versions of this product

Catalog No. AA1052418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-56-3
    Physical Form Powder
    Assay Percent Range 98% min.
    Molecular Formula P2O5
    Formula Weight 141.94
    MDL Number MFCD00011440
    Synonym Phosphoric anhydride
    Chemical Name or Material Phosphorus(V) oxide
    Grade ACS Grade

    Specifications

    Physical Form Powder
    Formula Weight 141.94
    Quantity 50g
    Color White
    DOT Information DOT Class: 8, Packaging Group: II
    EINECS Number 215-236-1
    Odor Odorless
    Assay Percent Range 98% min.
    Molecular Formula P2O5
    Appearance White powder
    MDL Number MFCD00011440
    CAS 1314-56-3
    Synonym Phosphoric anhydride
    Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Phosphorus(V) oxide
    Grade ACS Grade

    H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit