Alfa Aesar™ Phosphorus(V) oxide, Puratronic™, 99.9925% (metals basis)

Manufacturer: Alfa Aesar™ 1082722

Quantity 100g

 View more versions of this product

Catalog No. AA1082722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-56-3
    Formula Weight 141.94
    MDL Number MFCD00011440
    UN Number UN1807
    Merck Index 14,7355
    Chemical Name or Material Phosphorus(V) oxide
    Grade Puratronic™

    Specifications

    Formula Weight 141.94
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011440
    UN Number UN1807
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    Melting Point 340°C
    Physical Form Powder
    CAS 1314-56-3
    Merck Index 14,7355
    Health Hazard 1 Danger
    Density 2.39g/mL
    Odor Weak
    Sensitivity Moisture Sensitive
    Boiling Point 360°C (subl.)
    TSCA TSCA
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHOSPHORUS PENTOXIDE
    Quantity 100g
    RTECS Number TH3945000
    Chemical Name or Material Phosphorus(V) oxide
    Assay 99.9925%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit