Phosphorus(V) oxybromide, 98% min, Alfa Aesar™

As a chemical intermediate

Manufacturer: Alfa Aesar 5613522

Quantity 100g

 View more versions of this product

Catalog No. AA5613522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-59-5
    Molecular Formula POBr3
    Formula Weight 286.73
    MDL Number MFCD00036298
    UN Number UN1939
    Merck Index 14,7348
    Synonym Phosphorus(V) tribromide oxide; Phosphoryl bromide
    Chemical Name or Material Phosphorus(V) oxybromide

    Specifications

    Physical Form Lumps
    Formula Weight 286.73
    Quantity 100g
    TSCA TSCA
    Molecular Formula POBr3
    MDL Number MFCD00036298
    CAS 7789-59-5
    UN Number UN1939
    Merck Index 14,7348
    Synonym Phosphorus(V) tribromide oxide; Phosphoryl bromide
    Solubility Information Decomposes in water. Soluble in ether, benzene, and sulfuric acid
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318;
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
    Do not breathe dust/fume/gas/mist/vapours/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHOSPHORUS OXYBROMIDE
    Density 2.822
    Sensitivity Moisture Sensitive
    Melting Point 56°C
    Boiling Point 189°C
    Chemical Name or Material Phosphorus(V) oxybromide
    Assay ≥98%

    GHS H Statement
    H314-H318;
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
    Do not breathe dust/fume/gas/mist/vapours/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit