Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phosphorus(V) oxychloride, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1184714

Quantity 25g

 View more versions of this product

Catalog No. AA1184714


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10025-87-3
153.33
UN1810
Phosphorus(V) trichloride oxide; Phosphoryl chloride
POCl3
MFCD00011443
14,7349
Phosphorus(V) oxychloride

Specifications

1.645g/mL
Moisture Sensitive
Colorless
25g
1.460
POCl3
MFCD00011443
14,7349
Phosphorus(V) oxychloride
Pungent
105.8°C
1.25°C
10025-87-3
Liquid
153.33
UN1810
Phosphorus(V) trichloride oxide; Phosphoryl chloride
99.999%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H330-H372-H314-H302
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHOSPHORUS OXYCHLORIDE

RTECSNumber : TH4897000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit