Learn More
Enzo Life SciencesSupplier Diversity Partner Phosphoserine polyclonal antibody (HRP conjugate) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIKAPST2103E
Host: Rabbit. Immunogen: Phosphoserine conjugated to KLH and phosvitin mixture. Species Reactivity: Species independent. Applications: ELISA, WB. Recommended dilutions: ELISA (0.5µg/ml). Western Blot (2µg/ml, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In HRP stabilization buffer.
Catalog No. 50-666-596