Learn More
Phosphotungstic acid hydrate, ACROS Organics™
Specifications
Quantity | 25g |
Linear Formula | 12WO3·xH2O |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether and alcohol |
Formula Weight | 2880.17 |
Packaging | Plastic bottle |
Solubility | Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether and alcohol |
Chemical Name or Material | Phosphotungstic acid hydrate, p.a. |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger