Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotungstic Acid, n-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
12067-99-1
P2O5·24WO3·nH2O
12067-99-1
P2O5·24WO3·nH2O

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 2.13kg
Quantity 500g
Packaging Poly Bottle
Physical Form Crystals
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Phosphotungstic Acid, n-Hydrate