Learn More
Enzo Life SciencesSupplier Diversity Partner Phosphotyrosine polyclonal antibody (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIKAPTK155E
Host: Rabbit. Immunogen: Phosphotyrosine conjugated to KLH. Species Reactivity: Species independent. Applications: ELISA, IHC, IP, WB. Recommended dilutions: ELISA (1:1,000). Immunohistochemistry (1:50). Immunoprecipitation (10µg/500µg sample). Western Blot (1:250). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In PBS, pH 7.0, containing 0.01% sodium azide.
Catalog No. 50-666-647