Learn More
Enzo Life SciencesSupplier Diversity Partner Phosphotyrosine polyclonal antibody (HRP conjugate) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIKAPTK1553E
Host: Rabbit. Immunogen: Phosphotyrosine conjugated to KLH. Species Reactivity: Species independent. Applications: ELISA, WB. Recommended dilutions: ELISA (1:1,000). Western Blot (1µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In HRP stabilizing buffer with 50% glycerol.
Catalog No. 50-666-646