Learn More
Phosphotyrosine Mouse anti-Chemical, FITC, Clone: PY20, Exalpha
Mouse Monoclonal Antibody
Manufacturer:  Exalpha Biologicals IncSupplier Diversity Partner X1017
Encompass
 View more versions of this product
Catalog No. 50-210-1866
Add to cart
Description
Description
Phosphotyrosine Monoclonal antibody specifically detects Phosphotyrosine in Chemical samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Specifications
Specifications
Phosphotyrosine
Monoclonal
0.5-2 mg/mL
20mM sodium phosphate with 150mM NaCl, 50% glycerol and 3mM sodium azide; pH
Phosphotyrosine; pTyr; pY
Hybridoma produced from Balb/C mice immunized with phosphotyrosine coupled to carrier protein.
Protein A/G
RUO
Antibody
Monoclonal
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
PY20
FITC
Liquid
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical