Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotyrosine Mouse anti-Chemical, FITC, Clone: PY20, Exalpha Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$193.44 - $265.36

Specifications

Antigen Phosphotyrosine
Clone PY20
Concentration 0.5-2 mg/mL
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-1866 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
X1017
100 μg
Each for $265.36
Add to cart
 
50-210-1867 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
X1017S
50 μg
Each for $193.44
Add to cart
 
Description

Description

Phosphotyrosine Monoclonal antibody specifically detects Phosphotyrosine in Chemical samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Specifications

Specifications

Phosphotyrosine
0.5-2 mg/mL
Monoclonal
Liquid
RUO
20mM sodium phosphate with 150mM NaCl, 50% glycerol and 3mM sodium azide; pH
Hybridoma produced from Balb/C mice immunized with phosphotyrosine coupled to carrier protein.
Monoclonal
Protein A/G
Antibody
PY20
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
FITC
Mouse
Chemical
Phosphotyrosine; pTyr; pY
IgG2b
Primary
-20° C, Avoid Freeze/Thaw Cycles
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit