Learn More
Phosphotyrosine Mouse, Clone: PY793, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB14628
Catalog No. 89-034-711
Add to cart
Description
Description
Mouse monoclonal antibody raised against phosphotyrosine.
  • High specificity
  • High reactivity
Specifications
Specifications
Phosphotyrosine
PY793
Mouse monoclonal antibody raised against phosphotyrosine.
In 10 mM PBS.
Mouse
IgG2b
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Phosphotyrosine conjugated to BSA.
Protein A/G purification
RUO
Primary
This antibody shows no cross-reaction with other phosphoamino acids.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)