Learn More
Phosphotyrosine Rabbit, Clone: RM111, Abnova™
Rabbit monoclonal antibody raised against phosphotyrosine.
Manufacturer:  Abnova Corporation MAB12826
Catalog No. 89-934-767
Add to cart
Description
Description
Specifications
Specifications
Phosphotyrosine
RM111
Rabbit monoclonal antibody raised against phosphotyrosine.
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Rabbit
IgG
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
ELISA Flow Cytometry Immunocytochemistry (1:200-1:500) Immunohistochemistry (1:200-1:500) Immunoprecipitation (1:200-1:1000) Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Liquid
Phosphotyrosine conjugated with BSA.
Protein A affinity purification
RUO
Primary
This antibody reacts to tyrosine-phosphorylated proteins. No cross reactivity with nonphosphorylated tyrosine, phosphoserine, and phosphorthreonine