Learn More
Tocris Bioscience™ Photo-lenalidomide-acid
Functionalized photoswitchable cereblon ligand for Degraders
Manufacturer:  Tocris Bioscience™ 7328/50
Catalog No. 73-285-0
Add to cart
This item is not returnable. View return policy
Description
Description
Photo-lenalidomide-acid is a functionalized cereblon ligand containing a dimethoxy azo-benzene photoswitch; for use as a precursor to photoswitchable Degraders (PHOTACs). Photo-lenalidomide-acid is supplied with a carboxylic acid functional handle for ready conjugation to a linker/target protein ligand. Part of a range of functionalized tool molecules for Degrader R&D.
Specifications
Specifications
C23H22N4O8
0.95
2363154-39-4
Degrader Building Blocks
(E)-2-(4-((2-(2,6-Dioxopiperidin-3-yl)-1-oxoisoindolin-4-yl)diazenyl)-2,6-dimethoxyphenoxy)acetic acid
50 mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C