Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHPT1 Rabbit anti-Human, Janelia Fluor 646, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP299999JF646
Description
PHPT1 Polyclonal antibody specifically detects PHPT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PHPT1 | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein janus-a | |
Produced in rabbits immunized with purified, recombinant PHPT1 (Q9NRX4-1; Ala2-Tyr125). | |
Affinity purified | |
RUO | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
Lipid and Metabolism | |
Primary | |
29085 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title