Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phthalamidoxime, 97%

Click to view available options
Quantity:
250mg
1g
885960-32-7
C8H10N4O2
194.194
CQHMIKPVOMZRKW-KQQUZDAGSA-N
1,2-bis carboxamidoximo benzene
6346926
N-[(E)-amino-[(6E)-6-[amino(nitroso)methylidene]cyclohexa-2,4-dien-1-ylidene]methyl]hydroxylamine
C1=CC(=C(N)NO)C(=C(N)N=O)C=C1
885960-32-7
C8H10N4O2
194.194
CQHMIKPVOMZRKW-KQQUZDAGSA-N
1,2-bis carboxamidoximo benzene
6346926
N-[(E)-amino-[(6E)-6-[amino(nitroso)methylidene]cyclohexa-2,4-dien-1-ylidene]methyl]hydroxylamine
C1=CC(=C(N)NO)C(=C(N)N=O)C=C1
Quantity 250mg
Color Cream
Formula Weight 194.19
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Phthalamidoxime