Alfa Aesar™ Phthalan, 98%

Manufacturer: Alfa Aesar™ A1021722

Quantity 100g

 View more versions of this product

Catalog No. AAA1021722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 496-14-0
    Molecular Formula C8H8O
    Formula Weight 120.15
    MDL Number MFCD00005932
    Beilstein 111932
    Synonym 1,3-Dihydrobenzo[c]furan; 1,3-Dihydroisobenzofuran
    Chemical Name or Material Phthalan

    Specifications

    Formula Weight 120.15
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H8O
    CAS 496-14-0
    Boiling Point 192°C
    Beilstein 111932
    Synonym 1,3-Dihydrobenzo[c]furan; 1,3-Dihydroisobenzofuran
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Flash Point 63°C (145°F)
    MDL Number MFCD00005932
    Quantity 100g
    Chemical Name or Material Phthalan
    Assay 98%

    Description

    Applications
    Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4 ft.-Di-tert-butyl­biphenyl­, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.

    Solubility
    Low aqueous solubility.

    Notes
    Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit