Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phthalein Complexon 98.0+%, TCI America™
Click to view available options
Quantity:
1 g
25 g
Specifications
Color | White-Yellow |
Quantity | 25 g |
Formula Weight | 636.61 |
Percent Purity | ≥98.0% (T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Phthalein Complexon [Chelating Indicator] |
Safety and Handling
TSCA : Yes