Phthalic acid, ACS, 99.5+%

Manufacturer: Alfa Aesar 4177022

Quantity 100g

 View more versions of this product

Catalog No. AA4177022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 88-99-3
    Molecular Formula C6H4(COOH)2
    Formula Weight 166.13
    MDL Number MFCD00002467
    Beilstein 608199
    Merck Index 14,7371
    Synonym Benzene-1, 2-dicarboxylic acid
    Chemical Name or Material Phthalic acid
    Grade For ACS analysis

    Specifications

    Formula Weight 166.13
    Quantity 100g
    TSCA TSCA
    MDL Number MFCD00002467
    Beilstein 608199
    Merck Index 14,7371
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water fo
    RTECS Number TH9625000
    Physical Form Powder
    CAS 88-99-3
    Assay Percent Range >99.5%
    Molecular Formula C6H4(COOH)2
    Synonym Benzene-1, 2-dicarboxylic acid
    Chemical Name or Material Phthalic acid
    Assay ≥99.5%
    Grade For ACS analysis

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water fo

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit