Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Phthalic anhydride, ACS, 99.0-100.2%

Manufacturer:  Alfa Aesar™ 4177157

Quantity 4 x 500g

 View more versions of this product

Catalog No. AA4177157


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

85-44-9
MFCD00005918
118515
Phthalic anhydride
148.12
UN2214
14,7372
ACS Grade

Specifications

1.53g/mL
Weak, characteristic
295°C (subl.)
4 x 500g
Flaky Crystals
MFCD00005918
118515
Phthalic anhydride
ACS Grade
152° (305°F)
Moisture Sensitive
131° to 133°C
85-44-9
148.12
UN2214
14,7372
99.0 to 100.2%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H334-H318-H302-H335-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PHTHALIC ANHYDRIDE

RTECSNumber : TI3150000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit