Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalimide 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
2.5kg
Packaging:
Plastic bottle
85-41-6
C8H5NO2
147.13
MFCD00005881
XKJCHHZQLQNZHY-UHFFFAOYSA-N
phthalimide,1h-isoindole-1,3 2h-dione,o-phthalimide,isoindoline-1,3-dione,o-phthalic imide,1,3-isoindolinedione,benzoimide,2-diazoindan-1,3-dione,phenylimide,phthalimid
6809
CHEBI:38817
isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)NC2=O
85-41-6
C8H5NO2
147.13
MFCD00005881
XKJCHHZQLQNZHY-UHFFFAOYSA-N
phthalimide,1h-isoindole-1,3 2h-dione,o-phthalimide,isoindoline-1,3-dione,o-phthalic imide,1,3-isoindolinedione,benzoimide,2-diazoindan-1,3-dione,phenylimide,phthalimid
6809
CHEBI:38817
isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)NC2=O
Flash Point 165°C
Boiling Point 366.0°C
Infrared Spectrum Authentic
Melting Point >233.5°C
Quantity 2.5kg
Sublimation Point 0366.0°C
Packaging Plastic bottle
Color White to Yellow
Assay Percent Range 99%
Beilstein 21, 458
Fieser 07,330
Merck Index 15, 7485
Solubility Information Solubility in water: 0.4-0.6g/L (20°C). Other solubilities: 5 g/100g boiling alcohol, freely soluble in aqueous alkali hydroxides, soluble in boiling acetic acid, acetone, hot ether, and dimethyl sulfoxide, almost insoluble in benzene, petroleum ether
Formula Weight 147.13
Physical Form Crystalline Flakes
Percent Purity 99%
Chemical Name or Material Phthalimide
Show More Show Less