Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalyl-DL-glutamic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
6349-98-0
C13H11NO6
277.23
MFCD00023059
FEFFSKLJNYRHQN-UHFFFAOYNA-N
Phthaloyl-DL-glutamic Acid
92225
2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)pentanedioic acid
OC(=O)CCC(N1C(=O)C2=CC=CC=C2C1=O)C(O)=O
6349-98-0
C13H11NO6
277.23
MFCD00023059
FEFFSKLJNYRHQN-UHFFFAOYNA-N
Phthaloyl-DL-glutamic Acid
92225
2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)pentanedioic acid
OC(=O)CCC(N1C(=O)C2=CC=CC=C2C1=O)C(O)=O
Color White
Melting Point 192°C
Quantity 25g
Formula Weight 277.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Phthalyl-DL-glutamic Acid

RTECSNumber : MA3810000

TSCA : Yes