Learn More
Thermo Scientific™ Phusion™ Blood Direct PCR Kit Promotion
Perfrom PCR directly from whole blood without DNA purification.
Manufacturer:  Thermo Scientific™ F547L
 View more versions of this product
Catalog No. F547LPM
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Phusion Blood Direct PCR Kit is designed for amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR. The modified Phusion Hot Start II High-Fidelity DNA Polymerase is resistant to PCR inhibitors present in blood and retains its activity even with samples containing 40% whole blood.
This extremely accurate proofreading DNA polymerase makes the Phusion Blood Direct PCR Kit an ideal choice even for demanding applications, such as cloning. Phusion Blood Direct PCR Kit also allows analysis of blood samples with different anticoagulants, storage conditions and from various species.
Highlights
  • No need for time-consuming and expensive DNA purification steps
  • Simple protocol requires minimal hands-on time
  • Extremely short PCR protocol times
  • Modified Phusion Hot Start II High-Fidelity DNA Polymerase delivers abundant yields of specific product
Specifications
Specifications
amplification of DNA directly from whole blood.
Phusion™ Blood Direct PCR Kit
Store at -20°C.
Phusion Blood DNA Polymerase, 2x Phusion Blood PCR Buffer (includes dNTPs and MgCl2), EDTA, MgCl2, DMSO, universal control primer mix
500 reactions