Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion Buffers

Choose between two buffer packs to optimize high-fidelity PCR or difficult or long template amplification using Thermo Scientific™ Phusion™ High-Fidelity DNA Polymerases.

Manufacturer:  Thermo Scientific™ F519S

 View more versions of this product

Catalog No. F519S

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases: 5X Phusion HF Buffer and 5X Phusion GC Buffer. The error rate of Phusion DNA Polymerase in HF Buffer (4.4 × 10-7) is lower than that in GC Buffer (9.5 × 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.

5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Detergent-free HF Buffer for Phusion DNA Polymerases Developed for DHPLC and microarray applications

Includes:

Detergent-free 5x Phusion HF Buffer, 50 mM MgCl2 solution, 100% DMSO.

Specifications

Specifications

Phusion™ GC Buffer Pack
5x Phusion GC Buffer, 50 mM MgCl2 solution, 100% DMSO.
Store at -20°C. Stable for one year from the packaging date.
Phusion DNA polymerases
3mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit