Learn More
Thermo Scientific™ Phusion Buffers
Choose between two buffer packs to optimize high-fidelity PCR or difficult or long template amplification using Thermo ScientificTM PhusionTM High-Fidelity DNA Polymerases.
Products 3
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
F520S
View Documents
Thermo Scientific™
F520S
Detergent-free Phusion™ HF Buffer Pack 4 x 1.5mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
F518S
View Documents
Thermo Scientific™
F518S
Phusion™ HF Buffer Pack 3mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
F519S
View Documents
Thermo Scientific™
F519S
Phusion™ GC Buffer Pack 3mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases: 5X Phusion HF Buffer and 5X Phusion GC Buffer. The error rate of Phusion DNA Polymerase in HF Buffer (4.4 × 10-7) is lower than that in GC Buffer (9.5 × 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.
5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
Detergent-free HF Buffer for Phusion DNA Polymerases Developed for DHPLC and microarray applications
Includes:
Detergent-free 5x Phusion HF Buffer, 50 mM MgCl2 solution, 100% DMSO.