Learn More
Thermo Scientific™ Phusion High-Fidelity DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Get superior performance for cloning and other applications requiring high fidelity with one of the most accurate thermostable DNA polymerases available.
Manufacturer:  Thermo Scientific™ F531L
 View more versions of this product
Catalog No. F531L
Add to cart
Description
Description
Phusion DNA Polymerases set a gold standard for PCR performance. They offer a combination of characteristics that no other enzyme can match. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion DNA Polymerase is the most accurate thermostable polymerase available. This feature makes Phusion DNA Polymerases superior choice for cloning and other applications requiring high fidelity. The unique structure of the enzyme enables short protocol times, abundant yields and robust performance, even in the presence of PCR inhibitors. For the researcher, that translates to ease-of-use and convenience. In addition, Phusion DNA Polymerases also produce higher yields with lower enzyme amounts than other DNA polymerases.
New Phusion Green format is a combination of Phusion DNA Polymerase and 5X Phusion Green Buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with excelent performance of Phusion DNA Polymerases and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
Phusion High-Fidelity PCR Master Mix with HF Buffer and Phusion High-Fidelity PCR Master Mix with GC Buffer are convenient 2x mixes containing Phusion DNA Polymerase, nucleotides and optimized HF or GC reaction buffer including MgCl2. Only template and primers need to be added by the user.
  • High fidelity (error rate 4.4 x 10-7 in Phusion HF Buffer)
  • Speed of the polymerase allows short extension times (15-30 s/kb)
  • Robust reactions, minimal optimization needed
  • Increased product yields with minimal enzyme amounts
Includes:
2x Phusion High-Fidelity PCR Master Mix with HF Buffer contains Phusion DNA Polymerase, 2x Phusion HF Buffer and 400µM of each dNTP. The master mix supplies 1.5 mM MgCl2 and 200µM dNTP in final reaction concentration. A separate tube of DMSO is provided with the product. 2x Phusion High-Fidelity PCR Master Mix with GC Buffer contains Phusion DNA Polymerase, 2x Phusion GC Buffer and 400µM of each dNTP. The master mix supplies 1.5 mM MgCl2 and 200µM dNTP in final reaction concentration. A separate tube of DMSO is provided with the product.
Recommended for:
Fast PCR, high-fidelity PCR, cloning, long or difficult amplicons, DHPLC, microarray
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Fast PCR, high-fidelity PCR, cloning, long or difficult amplicons, DHPLC, microarray
With High-Fidelity buffer
500 x 50uL Rxns
Recommended storage temperature -20°C. Stable for one year from the packaging date. After thawing the mix can be stored at +4°C for three months.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser»s own internal research. No other patent rights are conveyed expressly, by implication, or by estoppels. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.