Learn More
Thermo Scientific™ Phusion™ RT-PCR Kit
Optimize high-fidelity RT-PCR with this complete 2-step kit, including random primers, Oligo dT primer, and controls.
Manufacturer:  Thermo Scientific™ F546L
 View more versions of this product
Catalog No. F546L
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Phusion RT-PCR Kit is a complete 2-step kit designed for high-fidelity RT-PCR. The kit contains the full set of reagents required for performing cDNA synthesis and PCR in two steps.
In the first step, a variety of RNA templates and all priming options may be used for efficient synthesis of cDNA with M-MuLV Reverse Transcriptase. In the second step, Phusion Hot Start II High-Fidelity DNA Polymerase is used for cDNA amplification. This polymerase is extremely accurate and robust, and the hot start modification guarantees additional specificity during PCR, enabling accurate cDNA amplification with high yields and short cycling times. Phusion RT-PCR Kit is an ideal choice when producing cDNA for cloning and gene expression studies.
Phusion RT-PCR Kit contains all the necessary reagents for performing cDNA synthesis and DNA amplification. In the cDNA synthesis step, RNA is transcribed by Moloney Murine Leukemia Virus Reverse Transcriptase (M-MuLV RT). Random and oligo (dT) primers are included in the kit but gene-specific primers can also be used. In the PCR step, Phusion Hot Start High-Fidelity DNA Polymerase is used for cDNA amplification. The superior features of Phusion Hot Start DNA Polymerase ensure accurate and specific cDNA amplification with high yields and short cycling times.
  • High yields for a broad range of RT-PCR products
  • Accurate amplification of cDNA with Phusion Hot Start DNA Polymerase (52x Taq fidelity)
  • Short and simple cDNA synthesis and PCR protocols
  • Easy to use; the kit includes all necessary components for performing cDNA synthesis and PCR
Includes:
RT enzyme mix (M-MulV RNase H+ and RNase inhibitor), 10x RT Buffer (includes 50 mM MgCl2), Oligo(dT)15 primer, random primers (hexamers), dNTP mix, Phusion Hot Start II DNA Polymerase, 5x Phusion HF Buffer, control RNA with carrier, control primer mix.
Recommended for:
Detection of gene expression, generation of cDNA products with superior fidelity for cloning and sequencing
Specifications
Specifications
Detection of gene expression, generation of cDNA products with superior fidelity for cloning and sequencing
Phusion™ RT-PCR Kit
Store all kit components at -20°C. All the kit components can be refrozen and stored at -20°C without affecting the performance of the kit.
RT enzyme mix (M-MulV RNase H+ and RNase inhibitor), 10x RT Buffer (includes 50 mM MgCl2), Oligo(dT)15 primer, random primers (hexamers), dNTP mix, Phusion Hot Start II DNA Polymerase, 5x Phusion HF Buffer, control RNA with carrier, control primer mix.
100 reactions