Learn More
PHYH Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: S210-5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242201V
Catalog No. NBP242201V
Add to cart
Description
Description
PHYH Monoclonal antibody specifically detects PHYH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
PHYH
S210-5
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PHYH
Fusion protein amino acids 1-338 (full-length) of human PhyH. Rat: 78% identity (264/338 amino acids identical). Mouse: 78% identity (264/338 amino acids identical).
Protein G purified
Lipid and Metabolism, Vision
Primary
5264
Human, Mouse, Rat
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
LNAP1, PAHXLN1, peroxisomal, PhyH, PHYH1, phytanoyl-CoA 2-hydroxylase, phytanoyl-CoA hydroxylase, phytanoyl-CoA hydroxylase (Refsum disease), RD
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 40kDa.