Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHYH Mouse anti-Human, Mouse, Rat, FITC, Clone: S210-5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242201F

Catalog No. NBP242201F


Add to cart

Description

Description

PHYH Monoclonal antibody specifically detects PHYH in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

PHYH
S210-5
Immunocytochemistry/Immunofluorescence
Purified
PHYH
Fusion protein amino acids 1-338 (full-length) of human PhyH. Rat: 78% identity (264/338 amino acids identical). Mouse: 78% identity (264/338 amino acids identical).
Protein G purified
Lipid and Metabolism, Vision
Primary
5264
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
LNAP1, PAHXLN1, peroxisomal, PhyH, PHYH1, phytanoyl-CoA 2-hydroxylase, phytanoyl-CoA hydroxylase, phytanoyl-CoA hydroxylase (Refsum disease), RD
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 40kDa.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit