Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHYH Mouse anti-Human, Mouse, Rat, Clone: S210-5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527669

Catalog No. PIMA527669


Add to cart

Description

Description

PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.
Specifications

Specifications

PHYH
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
O14832, O35386, P57093
Phytanoyl-CoA dioxygenase, peroxisomal, PAHX, peroxisomal, PhyH, PHYH1, Phytanic acid oxidase, phytanoil-CoA alpha hydroxylase, phytanoyl CoA 2 hydroxylase, Phytanoyl CoA 2 oxoglutarate dioxygenase, Phytanoyl CoA alpha hydroxylase, Phytanoyl CoA dioxygenase, Phytanoyl CoA dioxygenase peroxisomal, Phytanoyl-CoA alpha-hydroxylase, Phytanoyl-CoA dioxygenase, RD, LN1, LNAP1, OTTHUMP00000019131, OTTHUMP00000019132, OTTHUMP00000179083, OTTHUMP00000216226,
Mouse
IgG1
100 μg
-20°C
Primary
114209, 16922, 5264
Immunocytochemistry, Immunofluorescence, Western Blot
S210-5
Unconjugated
PHYH
Liquid
PHYH
Fusion protein amino acids 1-338 (full-length) of human PhyH. Rat: 78% identity (264/338 amino acids identical). Mouse: 78% identity (264/338 amino acids identical).
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit