Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHYHD1 Polyclonal Antibody
PHYHD1 Polyclonal Antibody
Supplier: Thermo Scientific PA5110772
Description
PHYHD1 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
PHYHD1 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5SRE7 | |
PHYHD1 | |
Recombinant Protein corresonding to Human PHYHD1. Recombinant protein control fragment (Product #RP-89700). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
Unconjugated | |
PHYHD1 | |
Phyhd1; phytanoyl-CoA dioxygenase domain containing 1; phytanoyl-CoA dioxygenase domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254295 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction