Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHYHIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 169841AP

Catalog No. 50-173-1309


Add to cart

Description

Description

This antibody recognizes both PHYHIP (35-38 kDa) and PHYHIPL (43-45 kDa).

Its interaction with PHYH suggests a role in the development of the central system.
Specifications

Specifications

PHYHIP
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q568Z9, Q8K0S0, Q92561
DYRK1AP3, KIAA0273, PAHX AP, PAHX AP1, PAHXAP1, PHYHIP
Rabbit
IgG
150 μL
-20°C
Primary
105653, 290356, 9796
Western Blot
Unconjugated
PHYHIP
Liquid
Phyhip
PHYHIP Fusion Protein Ag10575
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit