Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phyproof™ (+)-Isocorydine hydrochloride, ≥98% (HPLC), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
10 mg
Specifications
Specifications
CAS | 13552-72-2 |
Product Line | phyproof |
Linear Formula | C20H23 NO4 · HCl |
MDL Number | MFCD00134198 |
Melting Point | 215°C to 218°C |
Synonym | 11-Hydroxy-1,2,10-trimethoxyaporphine; Isocorydine hydrochloride; Luteanine |
Type | Reference Substance |
Quantity | 10 mg |
Formula Weight | 377.86 |
Percent Purity | ≥98% (HPLC) |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction