Learn More
Selleck Chemical LLCSupplier Diversity Partner PI-103 10mg 371935-74-9  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S103810MG
PI-103 is a multi-targeted PI3K inhibitor for p110α/β/δ/γ with IC50 of 2 nM/3 nM/3 nM/15 nM in cell-free assays, less potent to mTOR/DNA-PK with IC50 of 30 nM/23 nM. PI-103 induces apoptosis in murine T-cell Lymphoma.
Catalog No. 50-700-5