Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PI 828, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
PI 3-Kinase inhibitor (IC50 values are 0.098, 0.183, 0.227 and 1.967 μM for p110β, p110α, p110δ and p110γ respectively) that displays higher potency than LY 294002 (Cat No. 1130). Can be immobilized onto a solid phase.
Specifications
Quantity | 10 mg |
Formula Weight | 322.36 |
Percent Purity | >99% |
Chemical Name or Material | PI 828 |
Safety and Handling
Recommended Storage : Store at +4°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title