Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva pI Markers PROMO

Cytiva pI Markers PROMO are lyophilized mixtures of stable, salt-free, highly purified proteins.

Specifications

Color White
Format Lyophilized
Quantity 10 Vials/ Kit
Storage Requirements 4°C
View More Specs

Products 2
Catalog Number Mfr. No. pH Range Price Quantity & Availability  
Catalog Number Mfr. No. pH Range Price Quantity & Availability  
45-000-081LR0
View Documents
Cytiva
29489319
3 to 10 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
45-000-083LR0
View Documents
Cytiva
29489320
5 to 10.5 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

The markers may be run in parallel with the experimental samples on isoelectric focusing gels and are not prestained.

  • The precisely defined pI of the markers allows a permanent and accurate calibration of sample components
  • pH gradients and pI can be measured to within 0.01 to 0.05 pH unit
  • The progress of the focusing experiment can be monitored visually by the focusing of methyl red or Cytochrome c

Not for use in denaturing (urea) systems.

Specifications

Specifications

White
Lyophilized
10 Vials/ Kit
4°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit