Learn More
Sigma Aldrich Fine Chemicals Biosciences Interferon gamma mouse mIf
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11276905001
Encompass
Recombinant Interferon-gamma mouse (mlfN-gamma) is produced in E. coli and purified by standard chromatographic techniques.Primary structure One polypeptide chain (131 amino acids) identical to mouse natural IfN- ? (133 amino acids) but lacking the last two amino acids at the carboxy-terminus and not glycosylated. Glycosylation is not essential for biological activity.
Catalog No. 50-290-5919
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.