Learn More
PI3K p110(alpha) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 218901AP
Catalog No. 50-173-3624
Add to cart
Description
Description
The antibody is specific to PIK3CA.
Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers.
Specifications
Specifications
PI3K p110(alpha)
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P42336, P42337
p110 alpha, PI3 kinase subunit alpha, PI3K, PI3K alpha, PI3K p110(alpha), PI3-kinase p110 subunit alpha, PIK3CA, PtdIns 3 kinase subunit alpha
Rabbit
IgG
150 μL
-20°C
Primary
170911, 18706, 5290
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
PIK3CA
Liquid
Pik3ca
PI3K p110(alpha) Fusion Protein Ag16586
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat