Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIK3R2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586770
Description
Purity is > 95% by SDS-PAGE.
Binds to activated (phosphorylated) protein-tyrosine kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane.
Specifications
PIK3R2 | |
Polyclonal | |
Unconjugated | |
PIK3R2 | |
MPPH; MPPH1; p85; P85B; p85beta; p85-BETA; phosphatidylinositol 3-kinase; phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; phosphatidylinositol 3-kinase regulartory subunit polypeptide 2; phosphatidylinositol 3-kinase regulatory subunit beta; phosphatidylinositol 3-kinase, regulartory subunit, polypeptide 2; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphoinositide-3-kinase regulatory subunit 2; phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 2 (beta); phosphoinositide-3-kinase, regulatory subunit 2 (p85 beta); phosphoinositide-3-kinase, regulatory subunit, polypeptide 2 (p85 beta); PI3K; PI3K regulatory subunit beta; PI3-kinase p85 subunit beta; PI3-kinase regulatory subunit beta; PI3-kinase subunit p85-beta; PIK3R2; ptdIns-3-kinase p85-beta; ptdIns-3-kinase regulatory subunit beta; ptdIns-3-kinase regulatory subunit p85-beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18709, 29741, 5296 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O00459, O08908, Q63788 | |
PIK3R2 | |
A synthetic peptide corresponding to residues in Human PI3K p85beta. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction