Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIAS1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | PIAS1 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB023262
|
Novus Biologicals
NBP294354100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB023263
|
Novus Biologicals
NBP29435420UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
PIAS1 Polyclonal antibody specifically detects PIAS1 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PIAS1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
AR interacting protein, DDXBP1MGC141878, DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1, DEAD/H box-binding protein 1, E3 SUMO-protein ligase PIAS1, GBPRNA helicase II-binding protein, GU/RH-II, Gu-binding protein, MGC141879, Protein inhibitor of activated STAT protein 1, protein inhibitor of activated STAT, 1, protein inhibitor of activated STAT-1, zinc finger, MIZ-type containing 3, ZMIZ3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 422-651 of human PIAS1 (NP_057250.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.3), 50% glycerol | |
8554 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title