Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piccolo Mouse anti-Human, DyLight™ 405, Clone: PCLO-01, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262209V

Catalog No. NB002490


Add to cart

Description

Description

Piccolo Monoclonal specifically detects Piccolo in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

Piccolo
PCLO-01
Western Blot, Flow Cytometry
Purified, Conjugated
PCLO
Human recombinant PCLO protein
Protein A purified
Store at 4°C in the dark.
Monoclonal
The antibody A53-B/A2 reacts with Rod domain of cytokeratin 19 (40 kDa) in human tissue. Cytokeratin 19 is not expressed in hepatocytes; it is often co-expressed with cytokeratin 7.
Western Blot, Flow Cytometry
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
piccolo (presynaptic cytomatrix protein), protein piccolo
Mouse
IgG1
0.1 mL
Primary
27445
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit