Learn More
Piccolo Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: 6H9-B6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP149453FR
Catalog No. NBP149453FR
Add to cart
Description
Description
Piccolo Monoclonal antibody specifically detects Piccolo in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Piccolo
6H9-B6
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
BC001304.1
piccolo (presynaptic cytomatrix protein), protein piccolo
Mouse
IgG2b Kappa
0.1 ml
Primary
27445
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Purified
PCLO
Full length protein, BC001304.1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 550kDa.