Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picolinic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
98-98-6
C6H5NO2
123.11
MFCD00006293
SIOXPEMLGUPBBT-UHFFFAOYSA-N
picolinic acid,2-pyridinecarboxylic acid,2-picolinic acid,2-carboxypyridine,alpha-picolinic acid,o-pyridinecarboxylic acid,pyridinecarboxylic acid,alpha-pyridinecarboxylic acid,picolinate,acide picolique
1018
CHEBI:28747
pyridine-2-carboxylic acid
C1=CC=NC(=C1)C(=O)O
98-98-6
C6H5NO2
123.11
MFCD00006293
SIOXPEMLGUPBBT-UHFFFAOYSA-N
picolinic acid,2-pyridinecarboxylic acid,2-picolinic acid,2-carboxypyridine,alpha-picolinic acid,o-pyridinecarboxylic acid,pyridinecarboxylic acid,alpha-pyridinecarboxylic acid,picolinate,acide picolique
1018
CHEBI:28747
pyridine-2-carboxylic acid
C1=CC=NC(=C1)C(=O)O
pH 3 to 4 (1% soln.)
Sulfated Ash 1% max.
Melting Point 136°C to 139°C
Infrared Spectrum Authentic
Water 0.5% max.
Color White
Quantity 2.5kg
Packaging Plastic bottle
Assay Percent Range 99%
Beilstein 22, 33
Fieser 05,538; 07,293
Merck Index 15, 7515
Solubility Information Solubility in water: 887g/L (20°C). Other solubilities: soluble in ethanol (63g/L at 20°C),in glacial,acetic acid;,practically insoluble in ether,chloroform and,carbon disulfide
Formula Weight 123.11
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Chemical Name or Material Picolinic acid, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed. Causes serious eye irritation.

GHS P Statement
Do not eat, drink or smoke when using this product. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling. Rinse mouth.

Warning

EINECSNumber : 202-719-7

RTECSNumber : TJ7344000

TSCA : TSCA