Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picolinic Acid N-oxide 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
824-40-8
C6H5NO3
139.11
MFCD00006198
FHYMLBVGNFVFBT-UHFFFAOYSA-N
picolinic acid n-oxide,2-carboxypyridin-1-ium-1-olate,pyridine-2-carboxylic acid 1-oxide,picolinic acid 1-oxide,2-carboxypyridine n-oxide,2-pyridinecarboxylic acid, 1-oxide,chembl73173,2-pyridinecarboxylicacid, 1-oxide,picolinate n-oxide,1-oxopicolinic acid
69986
1-oxidopyridin-1-ium-2-carboxylic acid
C1=CC=[N+](C(=C1)C(=O)O)[O-]
824-40-8
C6H5NO3
139.11
MFCD00006198
FHYMLBVGNFVFBT-UHFFFAOYSA-N
picolinic acid n-oxide,2-carboxypyridin-1-ium-1-olate,pyridine-2-carboxylic acid 1-oxide,picolinic acid 1-oxide,2-carboxypyridine n-oxide,2-pyridinecarboxylic acid, 1-oxide,chembl73173,2-pyridinecarboxylicacid, 1-oxide,picolinate n-oxide,1-oxopicolinic acid
69986
1-oxidopyridin-1-ium-2-carboxylic acid
C1=CC=[N+](C(=C1)C(=O)O)[O-]
Color White to Beige
Infrared Spectrum Authentic
Quantity 5g
Melting Point >150°C
Water 0.5% max. (K.F.)
Packaging Glass bottle
Assay Percent Range 97%
Formula Weight 139.11
Physical Form Crystals or Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Picolinic acid N-oxide, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 212-528-0