Alfa Aesar™ Picrolonic acid, 98+%

Manufacturer: Alfa Aesar™ A1083614

Quantity 25g

 View more versions of this product

Catalog No. AAA1083614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 550-74-3
    Molecular Formula C10H8N4O5
    Formula Weight 264.21
    MDL Number MFCD00003133
    Beilstein 623723
    Merck Index 14,7412
    Synonym 3-Methyl-4-nitro-1-(4-nitrophenyl)-2-pyrazolin-5-one; Picrolic acid
    Chemical Name or Material Picrolonic acid

    Specifications

    Formula Weight 264.21
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H341-H315-H319-H335
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 550-74-3
    Synonym 3-Methyl-4-nitro-1-(4-nitrophenyl)-2-pyrazolin-5-one; Picrolic acid
    Recommended Storage 2°C to 8°C
    Melting Point ≈120°C (dec.)
    TSCA TSCA
    Molecular Formula C10H8N4O5
    MDL Number MFCD00003133
    Merck Index 14,7412
    Health Hazard 1 Warning
    RTECS Number UQ9510000
    Quantity 25g
    Beilstein 623723
    Chemical Name or Material Picrolonic acid
    Assay ≥98%

    Description

    Applications
    Reagent for the isolation of amines as crystalline π-complexes.Picrolonic acid is an analytical reagent used for the estimation of calcium, magnesium, lead, strontium and thorium. It is also used for the analysis of alkaloids, tryptophan and phenylalanine.

    Solubility
    Slightly soluble in water. Soluble in alcohol.

    Notes
    Incompatible with strong oxidizing agents and strong bases.

    GHS H Statement
    H341-H315-H319-H335
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit