Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Picrolonic acid, 98+%

Manufacturer:  Alfa Aesar™ A1083622

Quantity 100g

 View more versions of this product

Catalog No. AAA1083622


  Check Availability
Add to cart

Specifications

Description

Applications
Reagent for the isolation of amines as crystalline π-complexes.Picrolonic acid is an analytical reagent used for the estimation of calcium, magnesium, lead, strontium and thorium. It is also used for the analysis of alkaloids, tryptophan and phenylalanine.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
Description & Specifications

Chemical Identifiers

550-74-3
264.21
623723
3-Methyl-4-nitro-1-(4-nitrophenyl)-2-pyrazolin-5-one; Picrolic acid
C10H8N4O5
MFCD00003133
14,7412
Picrolonic acid

Specifications

≈120°C (dec.)
550-74-3
264.21
623723
3-Methyl-4-nitro-1-(4-nitrophenyl)-2-pyrazolin-5-one; Picrolic acid
≥98%
100g
C10H8N4O5
MFCD00003133
14,7412
Picrolonic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UQ9510000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit