Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Picrotoxin, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
This item is not returnable. View return policy
124-87-8
C30H34O13
602.58
VJKUPQSHOVKBCO-DMRKGALJSA-N
picrotoxin
131846146
CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O
This item is not returnable. View return policy
124-87-8
C30H34O13
602.58
VJKUPQSHOVKBCO-DMRKGALJSA-N
picrotoxin
131846146
CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O
Color White
Quantity 5g
Melting Point 199.0°C to 202.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Rotation − 32.00
Assay Percent Range 98%
Merck Index 15,7527
Solubility Information Solubility in water: 1g/5mL (100°C). Other solubilities: soluble in boiling alcohol (1gr/3mL),dilute acids,and alkalies; sparingly soluble in ether and ccl4,readily soluble in strong ammonia water and in aq.,solns of naoh.,1gr/13.5 mL in 95% ethan
Formula Weight 602.58
Physical Form Crystals or Powder
Percent Purity 98%
Chemical Name or Material Picrotoxin
Show More Show Less

Recommended Storage : 4°C